3H-Imidazo[4,5-c]pyridine 98%
Supplier: Thermo Scientific
Synonyms:
5-Aza-1H-benzoimidazole, 5-Azabenzimidazole
L06930.03
L06930.MD
ALFAL06930.03EA
238
SGD
ALFAL06930.03
ALFAL06930.MD
3H-Imidazo[4,5-c]pyridine 98%
3H-Imidazo[4,5-c]pyridine
Purity: 98%
CAS No.: 272-97-9
Molecular Formula: C6H5N3
Formula Weight: 119.13
Melting Point: 170-173°
Beilstein Reference: 2601
MDL No.: MFCD00051808
Formula:
C₆H₅N₃ MW: 119,13 g/mol Melting Pt: 170…173 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051808 CAS Number: 272-97-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...