4-Amino-6-mercapto-1H-pyrazolo[3,4-d]pyrimidine 94%

Supplier: Thermo Scientific

L06934.03 L06934.MD
ALFAL06934.03EA 0 SGD
ALFAL06934.03 ALFAL06934.MD
4-Amino-6-mercapto-1H-pyrazolo[3,4-d]pyrimidine 94%
4-Amino-6-mercapto-1H-pyrazolo[3,4-d]pyrimidine

Purity: 94% CAS No.: 23771-52-0 Molecular Formula: C5H5N5S Formula Weight: 167.19 Melting Point: 300° Beilstein Reference: 152193 MDL No.: MFCD00005690

Formula: C₅H₅N₅S
MW: 167,19 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00005690
CAS Number: 23771-52-0
EINECS: 245-872-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR