Ethyl-4-amino-2-mercaptopyrimidine-5-carboxylate 97%
Supplier: Thermo Scientific
|
Warning
|
L07121.03
L07121.06
ALFAL07121.03EA
0
SGD
ALFAL07121.03
ALFAL07121.06
Ethyl-4-amino-2-mercaptopyrimidine-5-carboxylate 97%
Ethyl-4-amino-2-mercaptopyrimidine-5-carboxylate
Purity: 97%
CAS No.: 774-07-2
Molecular Formula: C7H9N3O2S
Formula Weight: 199.23
Melting Point: ca 260° dec.
Beilstein Reference: 176363
MDL No.: MFCD00023227
Formula:
C₇H₉N₃O₂S MW: 199,23 g/mol Melting Pt: ∼ 260 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00023227 CAS Number: 774-07-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...