Methanesulphonylacetic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-(Methylsulphonyl)acetic acid
L07598.03
L07598.06
ALFAL07598.03EA
0
SGD
ALFAL07598.03
ALFAL07598.06
Methanesulphonylacetic acid ≥98%
Methanesulphonylacetic acid
Purity: 98+%
CAS No.: 2516-97-4
Molecular Formula: C3H6O4S
Formula Weight: 138.14
Melting Point: 114-119°
Beilstein Reference: 1760342
MDL No.: MFCD00051843
Formula:
C₃H₆O₄S MW: 138,14 g/mol Melting Pt: 114…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051843 CAS Number: 2516-97-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...