(3-Methoxyphenyl)hydrazine hydrochloride 98%

Supplier: Thermo Scientific
Danger

Synonyms: 3-Methoxyphenylhydrazine hydrochloride

L08091.03 L08091.06
ALFAL08091.03EA 129 SGD
ALFAL08091.03 ALFAL08091.06
(3-Methoxyphenyl)hydrazine hydrochloride 98%
(3-Methoxyphenyl)hydrazine hydrochloride

Purity: 98% CAS No.: 39232-91-2 Molecular Formula: C7H10N2O.HCl Formula Weight: 174.63 Storage Temperature: -30°C to -10°C Melting Point: ca 142° dec. Beilstein Reference: 5304389 MDL No.: MFCD00044416

Formula: C₇H₁₁ClN₂O
MW: 174,63 g/mol
Melting Pt: ∼ 142 °C (dec.)
MDL Number: MFCD00044416
CAS Number: 39232-91-2
EINECS: 254-368-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR