N-(tert-Butoxycarbonyl)-L-methionine ≥98%
Supplier: Thermo Scientific
L08366.03
L08366.06
ALFAL08366.03EA
38.4
SGD
ALFAL08366.03
ALFAL08366.06
N-(tert-Butoxycarbonyl)-L-methionine ≥98%
N-(tert-Butoxycarbonyl)-L-methionine
Purity: 98+%
CAS No.: 2488-15-5
Molecular Formula: C10H19NO4S
Formula Weight: 249.32
Storage Temperature: 2°C to 8°C
Odour: Odourless
Melting Point: 48-51°
Beilstein Reference: 1727869
Optical Rotation: -22° (c=1 in methanol)
MDL No.: MFCD00065586
Formula:
C₁₀H₁₉NO₄S MW: 249,33 g/mol Melting Pt: 48…51 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00065586 CAS Number: 2488-15-5 EINECS: 219-639-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...