N-Carbobenzyloxy-D-alanine ≥98%

Supplier: Thermo Scientific

Synonyms: Z-D-Ala-OH, N-Cbz-D-alanine

L08382.03 L08382.06
ALFAL08382.03EA 0 SGD
ALFAL08382.03 ALFAL08382.06
N-Carbobenzyloxy-D-alanine ≥98%
N-Carbobenzyloxy-D-alanine

Purity: 98+% CAS No.: 26607-51-2 Molecular Formula: C11H13NO4 Formula Weight: 223.23 Melting Point: 83-86° Beilstein Reference: 2056163 Optical Rotation: +14.5° (c=2 in acetic acid) MDL No.: MFCD00063126

Formula: C₁₁H₁₃NO₄
MW: 223,2 g/mol
Melting Pt: 83…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00063126
CAS Number: 26607-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR