Benzo[b]thiophene-3-acetonitrile ≥98%

Supplier: Thermo Scientific
Danger

L08547.03 L08547.06
ALFAL08547.03EA 159 SGD
ALFAL08547.03 ALFAL08547.06
Benzo[b]thiophene-3-acetonitrile ≥98%
Benzo[b]thiophene-3-acetonitrile

Purity: 98+% CAS No.: 3216-48-6 Molecular Formula: C10H7NS Formula Weight: 173.24 Melting Point: 62-66° Beilstein Reference: 125487 UN No.: UN3439 MDL No.: MFCD00027389

Formula: C₁₀H₇NS
MW: 173,24 g/mol
Melting Pt: 62…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00027389
CAS Number: 3216-48-6
EINECS: 221-733-4
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR