β-Cholestanol 98%
Supplier: Thermo Scientific
Synonyms:
beta-Cholestanol, 5A-Cholestan-3B-Ol, 5-ɑ-Cholestan-3-β-ol, Dihydrocholesterol
L08624.06
L08624.14
ALFAL08624.06EA
145
SGD
ALFAL08624.06
ALFAL08624.14
β-Cholestanol 98%
beta-Cholestanol
Purity: 98%
CAS No.: 80-97-7
Molecular Formula: C27H48O
Formula Weight: 388.68
Melting Point: 140-144°
Beilstein Reference: 2418594
Optical Rotation: +24° (c=1 in chloroform)
MDL No.: MFCD00066413
Formula:
C₂₇H₄₈O MW: 388,68 g/mol Melting Pt: 140…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066413 CAS Number: 80-97-7 EINECS: 201-315-8 Merck Index: 14,2200 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...