Ethyl-2,4-dimethylnicotinate 96%
Supplier: Thermo Scientific
Synonyms:
Ethyl 2,4-dimethylpyridine-3-carboxylate, Ethyl 2,4-dimethylnicotinate
L08672.06
L08672.03
ALFAL08672.06EA
0
SGD
ALFAL08672.06
ALFAL08672.03
Ethyl-2,4-dimethylnicotinate 96%
Ethyl-2,4-dimethylnicotinate
Purity: 96%
CAS No.: 37669-78-6
Molecular Formula: C10H13NO2
Formula Weight: 179.22
Refractive Index: 1.4980
Boiling Point: 116-118°/10mm
Beilstein Reference: 133910
MDL No.: MFCD00051962
Formula:
C₁₀H₁₃NO₂ MW: 179,22 g/mol Boiling Pt: 116…118 °C (10 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00051962 CAS Number: 37669-78-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...