N-(tert-Butoxycarbonyl)-D-methionine ≥98%
Supplier: Thermo Scientific
Synonyms:
(R)-2-((tert-Butoxycarbonyl)amino)-4-(methylthio)butanoic acid, Boc-D-Met-OH, N-Boc-D-methionine
L09207.03
L09207.06
ALFAL09207.03EA
67.7
SGD
ALFAL09207.03
ALFAL09207.06
N-(tert-Butoxycarbonyl)-D-methionine ≥98%
N-(tert-Butoxycarbonyl)-D-methionine
Purity: 98+%
CAS No.: 5241-66-7
Molecular Formula: C10H19NO4S
Formula Weight: 249.32
Storage Temperature: 2°C to 8°C
Melting Point: 47-50°
Beilstein Reference: 4294122
Optical Rotation: +22° (c=1 in methanol)
MDL No.: MFCD00038256
Formula:
C₁₀H₁₉NO₄S MW: 249,33 g/mol Melting Pt: 47…50 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00038256 CAS Number: 5241-66-7 EINECS: 226-043-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...