2,3,4,5-Tetramethyl-2-cyclopentenone cis- and trans-mixture 95%

Supplier: Thermo Scientific
Warning

Synonyms: 2,3,4,5-Tetramethylcyclopent-2-enone

L09617.06 L09617.14
ALFAL09617.06EA 177 SGD
ALFAL09617.06 ALFAL09617.14
2,3,4,5-Tetramethyl-2-cyclopentenone cis- and trans-mixture 95%
2,3,4,5-Tetramethyl-2-cyclopentenone

Purity: 95% CAS No.: 54458-61-6 Molecular Formula: C9H14O Formula Weight: 138.21 Flash Point: 73°(163°F) Refractive Index: 1.4760 Boiling Point: 99-100°/30mm Beilstein Reference: 2324088 MDL No.: MFCD00010248

Formula: C₉H₁₄O
MW: 138,21 g/mol
Boiling Pt: 99…100 °C (30 mmHg)
Density: 0,917
Flash Pt: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010248
CAS Number: 54458-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR