4-Acetyl-4'-methoxydiphenyl ether 97%
Supplier: Thermo Scientific
Synonyms:
1-(4-(4-Methoxyphenoxy)phenyl)ethanone, 4'-(4-Methoxyphenoxy)acetophenone, 4-Acetyl-4'-methoxydiphenyl ether
L09829.03
L09829.06
ALFAL09829.03EA
0
SGD
ALFAL09829.03
ALFAL09829.06
4-Acetyl-4'-methoxydiphenyl ether 97%
4-Acetyl-4'-methoxydiphenyl ether
Purity: 97%
CAS No.: 54916-28-8
Molecular Formula: C15H14O3
Formula Weight: 242.28
Melting Point: 56-58°
Boiling Point: 198-201°/2mm
Beilstein Reference: 2115929
MDL No.: MFCD00052928
Formula:
C₁₅H₁₄O₃ MW: 242,28 g/mol Boiling Pt: 198…201 °C (2 mmHg) Melting Pt: 56…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052928 CAS Number: 54916-28-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...