Methyl-2,4-dimethoxy-6-methylbenzoate 98%
Supplier: Thermo Fisher Scientific
L09890.03
L09890.MD
ALFAL09890.03EA
234
SGD
ALFAL09890.03
ALFAL09890.MD
Methyl-2,4-dimethoxy-6-methylbenzoate 98%
Methyl-2,4-dimethoxy-6-methylbenzoate
Purity: 98%
CAS No.: 6110-37-8
Molecular Formula: C11H14O4
Formula Weight: 210.23
Melting Point: 48-50°
Beilstein Reference: 1372278
MDL No.: MFCD00082782
Formula:
C₁₁H₁₄O₄ MW: 210.23 g/mol Melting Pt: 48…50 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082782 CAS Number: 6110-37-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...