Potassium acrylate hydrate 95%

Supplier: Thermo Scientific
Warning

L10079.06 L10079.14
ALFAL10079.06EA 117 SGD
ALFAL10079.06 ALFAL10079.14
Potassium acrylate hydrate 95%
Potassium acrylate hydrate

Purity: 95% CAS No.: 10192-85-5 Molecular Formula: H2C=CHCO2K.xH2O Formula Weight: 110.16(anhy) Storage Temperature: 2°C to 8°C Melting Point: 300° Beilstein Reference: 3695913 MDL No.: MFCD00151663

Formula: H₂C=CHCO₂K·xH₂O
MW: 110,16 g/mol (anhy)
Melting Pt: >300 °C
MDL Number: MFCD00151663
CAS Number: 10192-85-5
EINECS: 233-473-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR