4-Fluoro-DL-glutamic acid erythro + threo 97%

Supplier: Thermo Scientific

L10517.MB L10517.MC
ALFAL10517.MBEA 93.8 SGD
ALFAL10517.MB ALFAL10517.MC
4-Fluoro-DL-glutamic acid erythro + threo 97%
4-Fluoro-DL-glutamic acid

Purity: 97% CAS No.: 2708-77-2 Molecular Formula: C5H8FNO4 Formula Weight: 165.13 Melting Point: ca 185° dec. Beilstein Reference: 1947477 MDL No.: MFCD00055778

Formula: C₅H₈FNO₄
MW: 165,12 g/mol
Melting Pt: ∼ 185 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00055778
CAS Number: 2708-77-2
EINECS: 220-302-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR