3-Amino-5-(2-furyl)-1H-pyrazole 97%

Supplier: Thermo Scientific
Warning

Synonyms: 5-(Fur-2-yl)-1H-pyrazol-3-amine, 5-(Furan-2-yl)-1H-pyrazol-3-amine

L11721.03 L11721.MD
ALFAL11721.03EA 253 SGD
ALFAL11721.03 ALFAL11721.MD
3-Amino-5-(2-furyl)-1H-pyrazole 97%
3-Amino-5-(2-furyl)-1H-pyrazole

Purity: 97% CAS No.: 96799-02-9 Molecular Formula: C7H7N3O Formula Weight: 149.15 Melting Point: 124-128° Beilstein Reference: 4399373 MDL No.: MFCD00082696

Formula: C₇H₇N₃O
MW: 149,15 g/mol
Melting Pt: 124…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00082696
CAS Number: 96799-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR