Benzyl butyl ether 97%

Supplier: Thermo Scientific

L11747.06 L11747.14
ALFAL11747.06EA 85.4 SGD
ALFAL11747.06 ALFAL11747.14
Benzyl butyl ether 97%
Benzyl butyl ether

Purity: 97% CAS No.: 588-67-0 Molecular Formula: C6H5CH2O(CH2)3CH3 Formula Weight: 164.25 Refractive Index: 1.4910 Boiling Point: 111-112°/23mm Beilstein Reference: 1935234 MDL No.: MFCD00039954

Formula: C₁₁H₁₆O
MW: 164,25 g/mol
Boiling Pt: 111…112 °C (23 mmHg)
Density: 0,920
Storage Temperature: Ambient
MDL Number: MFCD00039954
CAS Number: 588-67-0
EINECS: 209-626-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR