4-Methoxy-2,3,6-trimethylbenzenesulphonyl chloride 90%, tech.
Supplier: Thermo Scientific
|
Danger
|
L11829.03
L11829.06
ALFAL11829.03EA
141
SGD
ALFAL11829.03
ALFAL11829.06
4-Methoxy-2,3,6-trimethylbenzenesulphonyl chloride 90%, tech.
4-Methoxy-2,3,6-trimethylbenzenesulphonyl chloride
Purity: 90%
CAS No.: 80745-07-9
Molecular Formula: C10H13ClO3S
Formula Weight: 248.73
Storage Temperature: -30°C to -10°C
Melting Point: 56-59°
Beilstein Reference: 4867665
UN No.: UN3261
MDL No.: MFCD00038846
Formula:
C₁₀H₁₃ClO₃S MW: 248,73 g/mol Melting Pt: 56…59 °C Storage Temperature: Freezer |
MDL Number:
MFCD00038846 CAS Number: 80745-07-9 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...