Morpholin-4-yl(phenyl)acetonitrile ≥98%

Supplier: Thermo Scientific
Danger

L12370.06 L12370.14
ALFAL12370.06EA 115 SGD
ALFAL12370.06 ALFAL12370.14
Morpholin-4-yl(phenyl)acetonitrile ≥98%
Morpholin-4-yl(phenyl)acetonitrile

Purity: 98+% CAS No.: 15190-10-0 Molecular Formula: C12H14N2O Formula Weight: 202.26 Melting Point: 67-71° Beilstein Reference: 181704 UN No.: UN3439 MDL No.: MFCD00014619

Formula: C₁₂H₁₄N₂O
MW: 202,26 g/mol
Melting Pt: 67…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00014619
CAS Number: 15190-10-0
EINECS: 239-242-9
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR