1,4-Bis(1-hydroxycyclohexyl)-1,3-butadiyne ≥98%
Supplier: Thermo Scientific
L12492.06
L12492.14
ALFAL12492.06EA
0
SGD
ALFAL12492.06
ALFAL12492.14
1,4-Bis(1-hydroxycyclohexyl)-1,3-butadiyne ≥98%
1,4-Bis(1-hydroxycyclohexyl)-1,3-butadiyne
Purity: 98+%
CAS No.: 5768-10-5
Molecular Formula: C16H22O2
Formula Weight: 246.35
Melting Point: 174-175°
Beilstein Reference: 1246867
MDL No.: MFCD00021401
Formula:
C₁₆H₂₂O₂ MW: 246,35 g/mol Melting Pt: 174…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00021401 CAS Number: 5768-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...