α-Benzylsuccinic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Benzylsuccinic acid

L12846.03 L12846.MD
ALFAL12846.03EA 0 SGD
ALFAL12846.03 ALFAL12846.MD
α-Benzylsuccinic acid ≥98%
alpha-Benzylsuccinic acid

Purity: 98+% CAS No.: 884-33-3 Molecular Formula: C11H12O4 Formula Weight: 208.22 Melting Point: 161-163° Beilstein Reference: 1966188 MDL No.: MFCD00055798

Formula: C₁₁H₁₂O₄
MW: 208,22 g/mol
Melting Pt: 161…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00055798
CAS Number: 884-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR