Thiophene-2-carboxylic anhydride 95%
Supplier: Thermo Scientific
|
Warning
|
L12851.03
L12851.06
ALFAL12851.03EA
105
SGD
ALFAL12851.03
ALFAL12851.06
Thiophene-2-carboxylic anhydride 95%
Thiophene-2-carboxylic anhydride
Purity: 95%
CAS No.: 25569-97-5
Molecular Formula: C10H6O3S2
Formula Weight: 238.29
Melting Point: 58-61°
Beilstein Reference: 177049
MDL No.: MFCD00068089
Formula:
C₁₀H₆O₃S₂ MW: 238,29 g/mol Melting Pt: 58…61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068089 CAS Number: 25569-97-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...