Ethyl-2-isocyanato-4-(methylthio)butyrate 97%
Supplier: Thermo Scientific
|
Danger
|
L13059.09
L13059.04
ALFAL13059.09EA
0
SGD
ALFAL13059.09
ALFAL13059.04
Ethyl-2-isocyanato-4-(methylthio)butyrate 97%
Ethyl-2-isocyanato-4-(methylthio)butyrate
Purity: 97%
CAS No.: 64505-12-0
Molecular Formula: C8H13NO3S
Formula Weight: 203.26
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4780
Boiling Point: 155°/24mm
Beilstein Reference: 1710840
UN No.: UN2206
MDL No.: MFCD00078433
Formula:
C₈H₁₃NO₃S MW: 203,26 g/mol Boiling Pt: 155 °C (24 mmHg) |
MDL Number:
MFCD00078433 CAS Number: 64505-12-0 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...