(+/-)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione ≥98%
Supplier: Thermo Scientific
L13326.03
L13326.06
ALFAL13326.03EA
364
SGD
ALFAL13326.03
ALFAL13326.06
(+/-)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione ≥98%
(+/-)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione
Purity: 98+%
CAS No.: 20007-72-1
Molecular Formula: C11H14O2
Formula Weight: 178.23
Storage Temperature: 2°C to 8°C
Melting Point: 47-50°
Beilstein Reference: 3201286
MDL No.: MFCD00001687
Formula:
C₁₁H₁₄O₂ MW: 178,23 g/mol Melting Pt: 47…50 °C |
MDL Number:
MFCD00001687 CAS Number: 20007-72-1 EINECS: 243-463-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...