(S)-(+)-3,4,8,8α-Tetrahydro-8α-methyl-1,6(2H,7H)-naphthalenedione 99%
Supplier: Thermo Scientific
Synonyms:
(S)-8a-Methyl-3,4,8,8a-tetrahydronaphthalene-1,6(2H,7H)-dione
L13334.04
L13334.ME
ALFAL13334.04EA
756
SGD
ALFAL13334.04
ALFAL13334.ME
(S)-(+)-3,4,8,8α-Tetrahydro-8α-methyl-1,6(2H,7H)-naphthalenedione 99%
(S)-(+)-3,4,8,8α-Tetrahydro-8α-methyl-1,6(2H,7H)-naphthalenedione
Purity: 99%
CAS No.: 33878-99-8
Molecular Formula: C11H14O2
Formula Weight: 178.23
Storage Temperature: 2°C to 8°C
Melting Point: 48-52°
Beilstein Reference: 1569979
Optical Rotation: +100° (c=1 in benzene)
MDL No.: MFCD00213797
Formula:
C₁₁H₁₄O₂ MW: 178,231 g/mol Melting Pt: 48…52 °C |
MDL Number:
MFCD00213797 CAS Number: 33878-99-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...