1-(2-Methyl-5-phenylthien-3-yl)ethanone 98%

Supplier: Thermo Scientific

L13554.03 L13554.06
ALFAL13554.03EA 0 SGD
ALFAL13554.03 ALFAL13554.06
1-(2-Methyl-5-phenylthien-3-yl)ethanone 98%
1-(2-Methyl-5-phenylthien-3-yl)ethanone

Purity: 98% CAS No.: 40932-63-6 Molecular Formula: C13H12OS Formula Weight: 216.30 Melting Point: 67-69° Beilstein Reference: 8481276 MDL No.: MFCD00151790

Formula: C₁₃H₁₂OS
Melting Pt: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00151790
CAS Number: 40932-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR