2,3-Dihydro-5H-1,4-benzodioxepin-5-one 98%
Supplier: Thermo Scientific
L13582.03
L13582.06
ALFAL13582.03EA
183
SGD
ALFAL13582.03
ALFAL13582.06
2,3-Dihydro-5H-1,4-benzodioxepin-5-one 98%
2,3-Dihydro-5H-1,4-benzodioxepin-5-one
Purity: 98%
CAS No.: 22891-52-7
Molecular Formula: C9H8O3
Formula Weight: 164.16
Refractive Index: 1.5580
Melting Point: 29-31°
Boiling Point: 178-184°/15mm
Beilstein Reference: 4464
MDL No.: MFCD00236035
Formula:
C₉H₈O₃ MW: 164,16 g/mol Boiling Pt: 178…184 °C (15 mmHg) Melting Pt: 29…31 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236035 CAS Number: 22891-52-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...