2,2,2-Trichloro-1,1-bis(trifluoromethyl)ethanol 97%
Supplier: Thermo Scientific
|
Warning
|
L13792.03
L13792.06
ALFAL13792.03EA
0
SGD
ALFAL13792.03
ALFAL13792.06
2,2,2-Trichloro-1,1-bis(trifluoromethyl)ethanol 97%
2,2,2-Trichloro-1,1-bis(trifluoromethyl)ethanol
Purity: 97%
CAS No.: 7594-49-2
Molecular Formula: C4HCl3F6O
Formula Weight: 285.40
Storage Temperature: 2°C to 8°C
Refractive Index: 1.3890
Boiling Point: 139°
Beilstein Reference: 1841641
MDL No.: MFCD00236021
Formula:
C₄HCl₃F₆O MW: 285,40 g/mol Boiling Pt: 139 °C |
MDL Number:
MFCD00236021 CAS Number: 7594-49-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...