(R)-(-)-2-Methoxy-2-phenylethanol ≥98%
Supplier: Thermo Scientific
L14044.03
L14044.MD
ALFAL14044.03EA
0
SGD
ALFAL14044.03
ALFAL14044.MD
(R)-(-)-2-Methoxy-2-phenylethanol ≥98%
(R)-(-)-2-Methoxy-2-phenylethanol
Purity: 98+%
CAS No.: 17628-72-7
Molecular Formula: C9H12O2
Formula Weight: 152.19
Flash Point: 97°(206°F)
Refractive Index: 1.5200
Boiling Point: 65°/0.1mm
Beilstein Reference: 2437247
Optical Rotation: -133° (c=1 in acetone)
MDL No.: MFCD00011550
Formula:
C₉H₁₂O₂ MW: 152,19 g/mol Boiling Pt: 65 °C (0,1 mmHg) Density: 1,050 Flash Pt: 97 °C (206 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011550 CAS Number: 17628-72-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...