Tri-O-acetyl-D-glucal 98%
Supplier: Thermo Scientific
Synonyms:
(-)-Tri-O-acetyl-D-glucal, 1,5-Anhydro-2-deoxy-D-arabino-hex-1-enitol 3,4,6-Tri-O-acetyl Ether, (2R,3S,4R)-2-(Acetoxymethyl)-3,4-dihydro-2H-pyran-3,4-diyl diacetate
L14103.06
L14103.14
ALFAL14103.06EA
0
SGD
ALFAL14103.06
ALFAL14103.14
Tri-O-acetyl-D-glucal 98%
Tri-O-acetyl-D-glucal
Purity: 98%
CAS No.: 2873-29-2
Molecular Formula: C12H16O7
Formula Weight: 272.26
Melting Point: 52-55°
Beilstein Reference: 90781
Optical Rotation: -25° (c=1 in methanol)
MDL No.: MFCD00063253
Formula:
C₁₂H₁₆O₇ MW: 272,25 g/mol Melting Pt: 52…55 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063253 CAS Number: 2873-29-2 EINECS: 220-709-0 |
Learn more
![](/m-sg.vwr.com/en_SG/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...