Methyl-3-(3-pyridyl)propionate 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Methyl 3-(pyridin-3-yl)propanoate
L14590.03
L14590.06
ALFAL14590.03EA
102
SGD
ALFAL14590.03
ALFAL14590.06
Methyl-3-(3-pyridyl)propionate 98%
Methyl-3-(3-pyridyl)propionate
Purity: 98%
CAS No.: 84199-98-4
Molecular Formula: C9H11NO2
Formula Weight: 165.19
Refractive Index: 1.5010
Boiling Point: 134°/12mm
Beilstein Reference: 132852
MDL No.: MFCD01075660
Formula:
C₉H₁₁NO₂ MW: 165,192 g/mol Boiling Pt: 134 °C (12 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD01075660 CAS Number: 84199-98-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...