1-Benzyl-4-Boc-piperazine 99%
Supplier: Thermo Scientific
|
Warning
|
L14644.06
L14644.03
ALFAL14644.06EA
400
SGD
ALFAL14644.06
ALFAL14644.03
1-Benzyl-4-Boc-piperazine 99%
1-Benzyl-4-Boc-piperazine
Purity: 99%
CAS No.: 57260-70-5
Molecular Formula: C16H24N2O2
Formula Weight: 276.38
Melting Point: 71-73°
Beilstein Reference: 797033
MDL No.: MFCD00075603
Formula:
C₁₆H₂₄N₂O₂ MW: 276,38 g/mol Melting Pt: 71…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075603 CAS Number: 57260-70-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...