4-Ethoxy-3-nitropyridine hydrochloride 97%
Supplier: Thermo Scientific
|
Warning
|
L14743.06
L14743.14
ALFAL14743.06EA
0
SGD
ALFAL14743.06
ALFAL14743.14
4-Ethoxy-3-nitropyridine hydrochloride 97%
4-Ethoxy-3-nitropyridine hydrochloride
Purity: 97%
CAS No.: 94602-04-7
Molecular Formula: C7H8N2O3.HCl
Formula Weight: 204.61
Melting Point: ca 270°
Beilstein Reference: 3712456
MDL No.: MFCD01075663
Formula:
C₇H₉ClN₂O₃ Melting Pt: ∼ 270 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075663 CAS Number: 94602-04-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...