Ethyl-5-acetyl-2-ethoxybenzoate 98%

Supplier: Thermo Scientific

L14750.14 L14750.22
ALFAL14750.14EA 209 SGD
ALFAL14750.14 ALFAL14750.22
Ethyl-5-acetyl-2-ethoxybenzoate 98%
Ethyl-5-acetyl-2-ethoxybenzoate

Purity: 98% CAS No.: 216143-90-7 Molecular Formula: C13H16O4 Formula Weight: 236.27 Melting Point: 51-54° MDL No.: MFCD01075665

Formula: C₁₃H₁₆O₄
MW: 236,27 g/mol
Melting Pt: 51…54 °C
Storage Temperature: Ambient
MDL Number: MFCD01075665
CAS Number: 216143-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR