3-Carboxy-2,2,5,5-tetramethylpyrrolidine-1-oxyl ≥97% free radical

Supplier: Thermo Scientific
Warning

Synonyms: 3-Carboxy-2,2,5,5-tetramethyl-1-pyrrolidinyloxy, 3-Carboxy-PROXYL

L15068.MF L15068.MD
ALFAL15068.MFEA 91.8 SGD
ALFAL15068.MF ALFAL15068.MD
3-Carboxy-2,2,5,5-tetramethylpyrrolidine-1-oxyl ≥97% free radical
3-Carboxy-2,2,5,5-tetramethylpyrrolidine-1-oxyl

Purity: 98% CAS No.: 2154-68-9 Molecular Formula: C9H16NO3 Formula Weight: 186.23 Melting Point: ca 200° dec. Beilstein Reference: 4136411 MDL No.: MFCD00003167

Formula: C₉H₁₆NO₃
MW: 186,23 g/mol
Melting Pt: ∼ 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003167
CAS Number: 2154-68-9
EINECS: 218-448-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR