Diethyl-2-(aminomethylene)malonate 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Diethyl 2-(aminomethylene)malonate
L15511.03
L15511.06
ALFAL15511.03EA
0
SGD
ALFAL15511.03
ALFAL15511.06
Diethyl-2-(aminomethylene)malonate 98%
Diethyl-2-(aminomethylene)malonate
Purity: 98%
CAS No.: 6296-99-7
Molecular Formula: C8H13NO4
Formula Weight: 187.20
Melting Point: 60-65°
Boiling Point: 114°/1mm
Beilstein Reference: 1708651
MDL No.: MFCD01075695
Formula:
C₈H₁₃NO₄ MW: 187,20 g/mol Boiling Pt: 114 °C (1 mmHg) Melting Pt: 60…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075695 CAS Number: 6296-99-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...