6,6-Dimethyl-5,7-dioxaspiro[2.5]octane-4,8-dione 99%
Supplier: Thermo Scientific
L15571.03
L15571.06
ALFAL15571.03EA
0
SGD
ALFAL15571.03
ALFAL15571.06
6,6-Dimethyl-5,7-dioxaspiro[2.5]octane-4,8-dione 99%
6,6-Dimethyl-5,7-dioxaspiro[2.5]octane-4,8-dione
Purity: 99%
CAS No.: 5617-70-9
Molecular Formula: C8H10O4
Formula Weight: 170.17
Melting Point: 63-66°
Beilstein Reference: 1241835
MDL No.: MFCD00042796
Formula:
C₈H₁₀O₄ Melting Pt: 63…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042796 CAS Number: 5617-70-9 EINECS: 227-044-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...