Ethyl-4-amino-1-piperidinecarboxylate 98%

Supplier: Thermo Scientific
Warning

Synonyms: Ethyl 4-amino-1-piperidinecarboxylate, 4-Amino-1-piperidinecarboxylic Acid Ethyl Ester

L15866.06 L15866.14
ALFAL15866.06EA 77.5 SGD
ALFAL15866.06 ALFAL15866.14
Ethyl-4-amino-1-piperidinecarboxylate 98%
Ethyl-4-amino-1-piperidinecarboxylate

Purity: 98% CAS No.: 58859-46-4 Molecular Formula: C8H16N2O2 Formula Weight: 172.23 Flash Point: 78°(172°F) Refractive Index: 1.4830 Boiling Point: 118-120°/6mm Beilstein Reference: 388597 MDL No.: MFCD00006484

Formula: C₈H₁₆N₂O₂
MW: 172,23 g/mol
Boiling Pt: 118…120 °C (6 mmHg)
Density: 1,004
Flash Pt: 78 °C (172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006484
CAS Number: 58859-46-4
EINECS: 261-472-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR