2,3-O-Isopropylidene-α,β-D-ribofuranose
Supplier: Thermo Scientific
Synonyms:
(4R,5R)-5-((R)-1,2-Dihydroxyethyl)-2,2-dimethyl-1,3-dioxolane-4-carbaldehyde
L15988.03
L15988.06
ALFAL15988.03EA
145
SGD
ALFAL15988.03
ALFAL15988.06
2,3-O-Isopropylidene-α,β-D-ribofuranose
2,3-O-Isopropylidene-alpha,beta-D-ribofuranose
CAS No.: 13199-25-2
Molecular Formula: C8H14O5
Formula Weight: 190.20
Storage Temperature: 2°C to 8°C
Optical Rotation: -27° (c=1 in chloroform)
MDL No.: MFCD01075718
Formula:
C₈H₁₄O₅ MW: 190,20 g/mol |
MDL Number:
MFCD01075718 CAS Number: 13199-25-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...