3-Bromothiophene-2-carboxamide 99%
Supplier: Thermo Scientific
|
Warning
|
L16138.03
L16138.MD
ALFAL16138.03EA
364
SGD
ALFAL16138.03
ALFAL16138.MD
3-Bromothiophene-2-carboxamide 99%
3-Bromothiophene-2-carboxamide
Purity: 99%
CAS No.: 78031-18-2
Molecular Formula: C5H4BrNOS
Formula Weight: 206.06
Melting Point: 106-108°
Beilstein Reference: 4937660
MDL No.: MFCD00173835
Formula:
C₅H₄BrNOS MW: 206,06 g/mol Melting Pt: 106…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00173835 CAS Number: 78031-18-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...