N-(4-Bromobutoxy)phthalimide ≥98%

Supplier: Thermo Fisher Scientific
Warning

L16314.03 L16314.06
ALFAL16314.03EA 0 SGD
ALFAL16314.03 ALFAL16314.06
N-(4-Bromobutoxy)phthalimide ≥98%
N-(4-Bromobutoxy)phthalimide

Purity: 98+% CAS No.: 5093-32-3 Molecular Formula: C12H12BrNO3 Formula Weight: 298.14 Melting Point: 70-72° Beilstein Reference: 1533519 MDL No.: MFCD00517028

Formula: C₁₂H₁₂BrNO₃
MW: 298.14 g/mol
Melting Pt: 70…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00517028
CAS Number: 5093-32-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR