(R)-(-)-2-Amino-3-methylbutane 98%, ee 97%, ChiPros®

Supplier: Thermo Scientific
Danger

L16330.06 L16330.03
ALFAL16330.06EA 434 SGD
ALFAL16330.06 ALFAL16330.03
(R)-(-)-2-Amino-3-methylbutane 98%, ee 97%, ChiPros®
(R)-(-)-2-Amino-3-methylbutane

Purity: 98% CAS No.: 34701-33-2 Molecular Formula: C5H13N Formula Weight: 87.16 Flash Point: 21°(69°F) Refractive Index: 1.4060 Boiling Point: 85-87° Beilstein Reference: 1718798 Optical Rotation: -5.2° (neat) UN No.: UN2733 MDL No.: MFCD01075731

Formula: C₅H₁₃N
MW: 87,16 g/mol
Boiling Pt: 85…87 °C
Density: 0,746
Flash Pt: <21 °C (69 °F)
Storage Temperature: Ambient
MDL Number: MFCD01075731
CAS Number: 34701-33-2
UN: 2733
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR