5-(5-Bromo-2-thienyl)-1H-tetrazole ≥98%

Supplier: Thermo Scientific
Warning

L16345.03 L16345.MD
ALFAL16345.03EA 0 SGD
ALFAL16345.03 ALFAL16345.MD
5-(5-Bromo-2-thienyl)-1H-tetrazole ≥98%
5-(5-Bromo-2-thienyl)-1H-tetrazole

Purity: 98+% CAS No.: 211943-12-3 Molecular Formula: C5H3BrN4S Formula Weight: 231.07 Melting Point: ca 225° dec. MDL No.: MFCD01075734

Formula: C₅H₃BrN₄S
MW: 231,07 g/mol
Melting Pt: ∼ 225 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01075734
CAS Number: 211943-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR