Bis(perfluoroisopropyl)ketone 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Bis(perfluoroisopropyl)methanone

L16708.03 L16708.06
ALFAL16708.03EA 83.8 SGD
ALFAL16708.03 ALFAL16708.06
Bis(perfluoroisopropyl)ketone 97%
Bis(perfluoroisopropyl)ketone

Purity: 97% CAS No.: 813-44-5 Molecular Formula: C7F14O Formula Weight: 366.06 Boiling Point: 72-73° Beilstein Reference: 1916532 UN No.: UN2810 MDL No.: MFCD00042087

Formula: C₇F₁₄O
MW: 366.06 g/mol
Boiling Pt: 72…73 °C
Density: 1.661
Storage Temperature: Ambient
MDL Number: MFCD00042087
CAS Number: 813-44-5
EINECS: 212-384-9
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR