Pyridine-3-boronic acid 1,3-propanediol ester ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-(1,3,2-Dioxaborinan-2-yl)pyridine
L17010.03
L17010.06
ALFAL17010.03EA
0
SGD
ALFAL17010.03
ALFAL17010.06
Pyridine-3-boronic acid 1,3-propanediol ester ≥98%
Pyridine-3-boronic acid 1,3-propanediol ester
Purity: 98+%
CAS No.: 131534-65-1
Molecular Formula: C8H10BNO2
Formula Weight: 162.98
Melting Point: 100-104°
Beilstein Reference: 6138748
MDL No.: MFCD01075749
Formula:
C₈H₁₀BNO₂ MW: 162,98 g/mol Melting Pt: 100…104 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075749 CAS Number: 131534-65-1 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...