(1R,2R)-(-)-2-Benzyloxycyclopentylamine ≥99%, ee 98%, ChiPros®

Supplier: Thermo Scientific
Danger

L17017.03 L17017.06
ALFAL17017.03EA 102 SGD
ALFAL17017.03 ALFAL17017.06
(1R,2R)-(-)-2-Benzyloxycyclopentylamine ≥99%, ee 98%, ChiPros®
(1R,2R)-(-)-2-Benzyloxycyclopentylamine

Purity: 99+% CAS No.: 181657-56-7 Molecular Formula: C12H17NO Formula Weight: 191.27 Flash Point: 110°(230°F) Refractive Index: 1.5305 Melting Point: 23° Boiling Point: 90-93°/0.23mm Optical Rotation: -57.5° (neat) UN No.: UN2735 MDL No.: MFCD01075750

Formula: C₁₂H₁₇NO
MW: 191,27 g/mol
Boiling Pt: 90…93 °C (0,23 mmHg)
Melting Pt: 23 °C
Density: 0,983
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01075750
CAS Number: 181657-56-7
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR