4-Benzyloxyindole-3-carbaldehyde 98%
Supplier: Thermo Scientific
L17567.03
L17567.MD
ALFAL17567.03EA
358
SGD
ALFAL17567.03
ALFAL17567.MD
4-Benzyloxyindole-3-carbaldehyde 98%
4-Benzyloxyindole-3-carbaldehyde
Purity: 98%
CAS No.: 7042-71-9
Molecular Formula: C16H13NO2
Formula Weight: 251.28
Melting Point: 167° dec.
Beilstein Reference: 1537328
MDL No.: MFCD00152002
Formula:
C₁₆H₁₃NO₂ MW: 251,28 g/mol Melting Pt: 167 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00152002 CAS Number: 7042-71-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...