DL-2-Amino-4-phosphonobutyric acid 95%
Supplier: Thermo Scientific
|
Warning
|
L17671.03
L17671.MD
ALFAL17671.03EA
0
SGD
ALFAL17671.03
ALFAL17671.MD
DL-2-Amino-4-phosphonobutyric acid 95%
DL-2-Amino-4-phosphonobutyric acid
Purity: 95%
CAS No.: 20263-07-4
Molecular Formula: C4H10NO5P
Formula Weight: 183.10
Melting Point: 225-228°
Beilstein Reference: 2093908
MDL No.: MFCD00013999
Formula:
C₄H₁₀NO₅P MW: 183,10 g/mol Melting Pt: 225…228 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013999 CAS Number: 20263-07-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...