4-(2-Pyridyl)-2-methyl-3-butyn-2-ol ≥98%
Supplier: Thermo Scientific
|
Warning
|
L17676.03
L17676.06
ALFAL17676.03EA
131
SGD
ALFAL17676.03
ALFAL17676.06
4-(2-Pyridyl)-2-methyl-3-butyn-2-ol ≥98%
4-(2-Pyridyl)-2-methyl-3-butyn-2-ol
Purity: 98+%
CAS No.: 29767-97-3
Molecular Formula: C10H11NO
Formula Weight: 161.20
Melting Point: 80-85°
Boiling Point: 120°/1.5mm
Beilstein Reference: 1447913
MDL No.: MFCD00168867
Formula:
C₁₀H₁₁NO MW: 161,20 g/mol Boiling Pt: 120 °C (1,5 mmHg) Melting Pt: 80…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00168867 CAS Number: 29767-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...